missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PRTFDC1 Polyclonal antibody specifically detects PRTFDC1 in Human, Rat samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | PRTFDC1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.4.2.8, FLJ11888, HHGPEC 2.4.2, phosphoribosyl transferase domain containing 1, phosphoribosyltransferase domain-containing protein 1 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 14 amino acid synthetic peptide from near the center of human PRTFDC1. The immunogen is located within amino acids 90 - 140 of PRTFDC1. Amino Acid Squence: MKVDFIRLKSYRND |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?