missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PRTFDC1 Monoclonal specifically detects PRTFDC1 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | PRTFDC1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1 |
| Conjugate | Biotin |
| Dilution | ELISA |
| Formulation | PBS |
| Gene Alias | EC 2.4.2.8, FLJ11888, HHGPEC 2.4.2, phosphoribosyl transferase domain containing 1, phosphoribosyltransferase domain-containing protein 1 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human PRTFDC1 (Q9NRG1-1; Met1-Val225). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?