missing translation for 'onlineSavingsMsg'
Learn More

PRT4165, MedChemExpress

Product Code. 16490388
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490388 5 mg 5mg
16487726 100 mg 100mg
16447928 10 mg 10mg
16443867 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490388 Supplier Medchem Express Supplier No. HY19817
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PRT4165 is a potent inhibitor of PRC1-mediated H2A ubiquitylation.

PRT4165 is a potent inhibitor of PRC1-mediated H2A ubiquitylation.

Specifications

Chemical Name or Material PRT4165
CAS 31083-55-3
Color Green
Purity Grade Notes Research
Molecular Formula C15H9NO2
Quantity 5 mg
Synonym NSC600157
Solubility Information DMSO : 25 mg/mL (106.27 mM; Need ultrasonic)
SMILES O=C1/C(C(C2=C1C=CC=C2)=O)=C/C3=CC=CN=C3
Molecular Weight (g/mol) 235.24
Formula Weight 235.24
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.