missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PRT 4165

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PRT 4165 is an inhibitor of Bmi1/Ring1A, subunits of the polycomb repressive complex 1 (PRC1); inhibits self-ubiquitination (IC50 = 3.9 μM) but does not increase cellular levels of either subunit. Prevents Bmi1/Ring1A-mediated ubiquitination and drug-induced degradation of topoisomerase 2α (Top2α). Also shown to inhibit the in vitro E3 ubiquitin ligase activity of RNF2 and a Bmi1/RNF2 complex, inhibiting H2A/H2AX ubiquitination. Blocks polycomb repressor complex (PRC) 1-mediated histone H2A ubiquitination in vitro.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | PRT 4165 |
| Purity | >99% |
| Molecular Weight (g/mol) | 235.24 |
| Inhibitors | PRT 4165 |
For Research Use Only
Product Content Correction
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu