missing translation for 'onlineSavingsMsg'
Learn More

PRT-060318, MedChemExpress

Product Code. 16475458
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475458 10 mg 10mg
16453346 25 mg 25mg
16441637 50 mg 50mg
16418177 5 mg 5mg
16406056 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16475458 Supplier Medchem Express Supplier No. HY12974

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PRT-060318 (PRT318) is a novel selective inhibitor of the tyrosine kinase Syk with an IC50 of 4 nM.

PRT-060318 (PRT318) is a novel selective inhibitor of the tyrosine kinase Syk with an IC50 of 4 nM.

Specifications

Chemical Name or Material PRT-060318
CAS 1194961-19-7
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C18H24N6O
Quantity 10 mg
Synonym PRT318
Solubility Information H2O : 5.6 mg/mL (16.45 mM; Need ultrasonic and warming)
SMILES O=C(C1=CN=C(N[C@H]2[C@@H](N)CCCC2)N=C1NC3=CC=CC(C)=C3)N
Molecular Weight (g/mol) 340.42
Formula Weight 340.42
Percent Purity 99.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.