missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRSS54 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572124
This item is not returnable.
View return policy
Description
PRSS54 Polyclonal Antibody for Western Blot
This gene encodes a putative serine-type endopeptidase containing the peptidase S1 domain. Alternative splicing of this gene results in multiple transcript variants.
Specifications
| PRSS54 | |
| Polyclonal | |
| Unconjugated | |
| PRSS54 | |
| 4931432M23Rik; Cancer/testis antigen 67; CT67; Inactive serine protease 54; Klkbl4; Plasma kallikrein-like protein 4; protease, serine 54; protease, serine, 54; PRSS54; testis tissue sperm-binding protein Li 40a | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 221191, 70993 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6PEW0, Q7M756 | |
| PRSS54 | |
| KLH conjugated synthetic peptide between 339-367 amino acids from the C-terminal region of human KLKBL4. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction