missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRSS53 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571247
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 92%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 79%; Rat: 93%.
In vitro can degrade the fibrinogen alpha chain of as well as pro-urokinase-type plasminogen activator.
Specifications
| PRSS53 | |
| Polyclonal | |
| Unconjugated | |
| PRSS53 | |
| EDTP308; mFLJ00289; POL3S; polyserase 3; polyserase-3; Polyserine protease 3; protease, serine 53; protease, serine, 53; PRSS53; RGD1311277; RGD1566127; Serine protease 53; UNQ308 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 339105 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q2L4Q9 | |
| PRSS53 | |
| synthetic peptide directed towards the N-terminal region of human PRSS53. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction