missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRSS38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551199
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total PRSS38 protein.
The function of this protein remains unknown.
Specifications
| PRSS38 | |
| Polyclonal | |
| Unconjugated | |
| PRSS38 | |
| marapsin 2; marapsin-2; MPN2; protease, serine 38; protease, serine, 38; PRSS38; Serine protease 38; serine protease MPN2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 339501 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| A1L453 | |
| PRSS38 | |
| Full length fusion protein of human PRSS38. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction