missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRSS38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563186
This item is not returnable.
View return policy
Description
Immunogen sequence: IYDMYVGLVN LRVAGNHTQW YEVNRVILHP TYEMYHPIGG DVALVQLKTR IVFSESVLPV CLATPE Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 64%.
The function of this protein remains unknown.
Specifications
| PRSS38 | |
| Polyclonal | |
| Unconjugated | |
| PRSS38 | |
| marapsin 2; marapsin-2; MPN2; protease, serine 38; protease, serine, 38; PRSS38; Serine protease 38; serine protease MPN2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 339501 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A1L453 | |
| PRSS38 | |
| Recombinant protein corresponding to Human PRSS38. Recombinant protein control fragment (Product #RP-103953). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction