missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRSS37 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554229
This item is not returnable.
View return policy
Description
Immunogen sequence: PQDDLMLIKL AKPAMLNPKV QPLTLATTNV RPGTVCLLSG LDWSQENSGR HPDLRQNLEA PVMSDREC Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.
PRSS37 plays a role in male fertility. May have a role in sperm migration or binding to zona-intact eggs. Involved in the activation of the proacrosin/acrosin system.
Specifications
| PRSS37 | |
| Polyclonal | |
| Unconjugated | |
| PRSS37 | |
| 1700016G05Rik; LOC540062 protein; Peptidase S1 domain-containing protein LOC136242; probable inactive serine protease 37; Probable inactive trypsin-X2; protease, serine 37; protease, serine, 37; Prss37; RGD1311823; serine protease 37; Tryx2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 136242 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A4D1T9 | |
| PRSS37 | |
| Recombinant protein corresponding to Human PRSS37. Recombinant protein control fragment (Product #RP-92860). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction