missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRSS35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558542
This item is not returnable.
View return policy
Description
Immunogen sequence: MEWDFMWHLR KVPRIVSERT FHLTSPAFEA DAKMMVNTVC GIECQKELPT PSLSELEDYL SYETVFENGT RTLTRVKVQD LVLEPTQNIT TKGVSVRRK Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 70%.
The PRSS35 (Protease, Serine 35) gene encodes a protein that belongs to the trypsin-like serine protease family, which is involved in various physiological processes, including digestion, immune response, and blood coagulation. PRSS35 is located on chromosome 6 in humans. The protein is believed to play a role in extracellular matrix remodeling and may be involved in processes such as tissue repair and inflammation. Expression of PRSS35 has been detected in multiple tissues, including the reproductive system, where it may influence fertility and reproductive tract function. However, the specific substrates and detailed biological functions of PRSS35 are still under investigation. Alterations in PRSS35 expression have been associated with certain pathological conditions, such as cancer and inflammatory diseases, suggesting its potential as a biomarker and therapeutic target.
Specifications
| PRSS35 | |
| Polyclonal | |
| Unconjugated | |
| PRSS35 | |
| 6030424L22Rik; C6orf158; dJ223E3.1; Inactive serine protease 35; P3D9; protease, serine 35; protease, serine, 35; PRSS35; UNQ522/PRO1057 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 167681 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N3Z0 | |
| PRSS35 | |
| Recombinant protein corresponding to Human PRSS35. Recombinant protein control fragment (Product #RP-94886). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction