Learn More
Invitrogen™ PRSS32 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547356
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human Tryptase-G1, recombinant mouse (rm) MCPT-6, and rmMCPT-7 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Specifications
| PRSS32 | |
| Polyclonal | |
| Unconjugated | |
| Prss32 | |
| 2010001P08Rik; eosinophil serine protease; mT5; protease, serine 32; protease, serine, 32; Prss32; testisin; tryptase 5; tryptase-5 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 69814 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| 0 | |
| Prss32 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Tryptase-5/Prss32 Ser20-Arg304. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.