missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRSS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598310
This item is not returnable.
View return policy
Description
PRSS3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
PRSS3 encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is expressed in the brain and pancreas and is resistant to common trypsin inhibitors. It is active on peptide linkages involving the carboxyl group of lysine or arginine. PRSS3 is localized to the locus of T cell receptor beta variable orphans on chromosome 9. Four transcript variants encoding different isoforms have been described for this gene.
Specifications
| PRSS3 | |
| Polyclonal | |
| Unconjugated | |
| PRSS3 | |
| Brain trypsinogen; mesotrypsin; Mesotrypsinogen; MTG; pancreatic trypsinogen III; protease, serine 3; protease, serine, 3; protease, serine, 4 (trypsin 4, brain); PRSS3; PRSS4; Serine protease 3; Serine protease 4; T9; Tb; Try3; TRY4; trypsin 3; trypsin III; Trypsin IV; trypsin-3; trypsinogen; trypsinogen 15; trypsinogen 4; trypsinogen 5; trypsinogen IV | |
| Rabbit | |
| Protein G | |
| RUO | |
| 5646 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P35030 | |
| PRSS3 | |
| Recombinant Human Trypsin-3 protein (81-303AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction