missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRSS3 Monoclonal Antibody (419911)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA524156
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
PRSS3 encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is expressed in the brain and pancreas and is resistant to common trypsin inhibitors. It is active on peptide linkages involving the carboxyl group of lysine or arginine. PRSS3 is localized to the locus of T cell receptor beta variable orphans on chromosome 9. Four transcript variants encoding different isoforms have been described for this gene.
Specifications
| PRSS3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| 0 | |
| PRSS3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Trypsin 3/PRSS3 Phe16-Asn246. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Frozen) | |
| 419911 | |
| Unconjugated | |
| PRSS3 | |
| Brain trypsinogen; mesotrypsin; Mesotrypsinogen; MTG; pancreatic trypsinogen III; protease, serine 3; protease, serine, 3; protease, serine, 4 (trypsin 4, brain); PRSS3; PRSS4; Serine protease 3; Serine protease 4; T9; Tb; Try3; TRY4; trypsin 3; trypsin III; Trypsin IV; trypsin-3; trypsinogen; trypsinogen 15; trypsinogen 4; trypsinogen 5; trypsinogen IV | |
| Rat | |
| Protein A/G | |
| RUO | |
| 22073 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction