missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRRX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120752
This item is not returnable.
View return policy
Description
Positive test controls include: NIH/3T3, Mouse brain, Mouse skin, Mouse spleen, Rat testis. Immunogen sequence: QEAAIEQPVA PRPTALSPDY LSWTASSPYS TVPPYSPGSS GPATPGVNMA NSIASLRLKA KEFSLHHSQV PTVN.
The paired-class homeobox genes PRX1 and PRX2 are necessary for craniofacial and limb development and are expressed in similar patterns in the cranial mesenchyme, limb buds, axial mesoderm and branchial arches. These proteins exhibit different patterns of expression, however, in heart and brain tissue. Specifically, PRX1 is expressed in the endocardial cusions, semilunar and atrioventricular valves, whereas PRX2 is initially expressed in a diffuse myocardial pattern and is later expressed in the ventricular septum. Furthermore, PRX2 is never expressed in the brain, whereas PRX1 is expressed in the ventral hypothalamus and in the telencephalon. Murine mutants lacking PRX1 function demonstrate skeletal defects in the skull, limbs, and vertebral column. Mice lacking functional PRX2 alone do not demonstrate skeletal abnormalities, however, PRX1/PRX2 double mutants demonstrate novel abnormalities that are not visualized with the PRX1-deficient mice. Transcripts of neither PRX1 nor PRX2 are detected in normal adult rat pulmonary arteries, however vascular disease induces PRX gene expression wherein they co-localize to sites of Tenascin-C expression.
Specifications
| PRRX2 | |
| Polyclonal | |
| Unconjugated | |
| Prrx2 | |
| homeo box of paired rule; homeobox protein S8; LOW QUALITY PROTEIN: paired mesoderm homeobox protein 2; MGC19843; paired mesoderm homeobox protein 2; paired related homeobox 2; paired-like homeodomain protein PRX2; paired-related homeobox protein 2; PMX2; PRRX2; Prx2; PRX-2; S8; surface antigen, homeo box of paired rule; testicular tissue protein Li 148; testicular tissue protein Li 160 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 113931, 20204, 51450 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.055 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q06348, Q99811 | |
| Prrx2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 180-253 of human PRRX2 (NP_057391.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction