missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PRRX1 Antibody (CL14471), Novus Biologicals™
Mouse Monoclonal Antibody
£336.00 - £538.00
Specifications
| Antigen | PRRX1 |
|---|---|
| Clone | CL14471 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL |
| Applications | Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043876
|
Novus Biologicals™
NBP3-26856-100ul |
100 μL |
£538.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043768
|
Novus Biologicals™
NBP3-26856-25ul |
25 μL |
£336.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRRX1 Monoclonal antibody specifically detects PRRX1 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry/ ImmunofluorescenceSpecifications
| PRRX1 | |
| 1 mg/mL | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Growth and Development, Neuronal Cell Markers | |
| PBS, pH 7.2, 40% glycerol | |
| 5396 | |
| IgG3 | |
| Protein A purified |
| CL14471 | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Homeobox protein PHOX1, paired mesoderm homeo box 1, paired mesoderm homeobox 1 isoform pmx-1b, paired mesoderm homeobox protein 1, paired related homeobox 1, Paired-related homeobox protein 1, PHOX1PRX-1, PMX1PRX1 | |
| This antibody was developed against a Synthetic Peptide of P54821. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title