missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRRX1 Antibody (CL14471) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26856AF532
This item is not returnable.
View return policy
Description
PRRX1 Monoclonal antibody specifically detects PRRX1 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry/ Immunofluorescence
Specifications
| PRRX1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| Homeobox protein PHOX1, paired mesoderm homeo box 1, paired mesoderm homeobox 1 isoform pmx-1b, paired mesoderm homeobox protein 1, paired related homeobox 1, Paired-related homeobox protein 1, PHOX1PRX-1, PMX1PRX1 | |
| This antibody was developed using a synthetic peptide derived from P54821, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers | |
| 5396 | |
| Store at 4⪚C in the dark. | |
| IgG3 |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CL14471 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction