missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| For Use With (Application) | Antibody Competition |
| Formulation | PBS and 1 M Urea, pH 7.4. |
| Gene ID (Entrez) | 80863 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 20 kDa |
| Quantity | 100 μL |
| Storage Requirements | Store at -20°C. Avoid freeze/thaw cycles. |
| Gene Alias | C6orf31, Chromosome 6 Open Reading Frame 31, Dispanin Subfamily D Member 1, DSPD1, IFITMD7, Interferon Induced Transmembrane Protein Domain Containing 7, NG5, Proline-Rich Transmembrane Protein 1 |
| Gene Symbol | PRRT1 |
| Inhibitor Properties | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-68749. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL |
| Product Line | Novus |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?