missing translation for 'onlineSavingsMsg'
Learn More

PRRSV nsp1 beta Antibody, Novus Biologicals™

Product Code. 18356395 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356395 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356395 Supplier Novus Biologicals Supplier No. NBP313276

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRRSV nsp1 beta Polyclonal antibody specifically detects PRRSV nsp1 beta in Virus samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRRSV nsp1 beta
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias Non-structural protein 1 beta, Nsp1-beta papain-like cysteine proteinase, PCP1-beta, Porcine reproductive and respiratory syndrome virus nsp1 beta
Host Species Rabbit
Immunogen Full length PRRSV nsp1 beta recombinant protein. (PRRS virus (strain VR-2332))
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.