missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRRSV GP3 Antibody, Novus Biologicals™
Description
PRRSV GP3 Polyclonal antibody specifically detects PRRSV GP3 in Virus samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PRRSV GP3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 20% Glycerol (pH7) |
| Gene Alias | Glycoprotein 3, Porcine reproductive and respiratory syndrome virus GP3, Protein GP3 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of PRRSV GP3 (PRRS virus (strain PrimePac PRRS vaccine)). The exact sequence is proprietary. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?