missing translation for 'onlineSavingsMsg'
Learn More

PRRG2 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£157.00 - £364.00

Specifications

Antigen PRRG2
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18657241
View Documents
Novus Biologicals
NBP2-93368-0.02ml
0.02 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18676891
View Documents
Novus Biologicals
NBP2-93368-0.1ml
0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

PRRG2 Polyclonal antibody specifically detects PRRG2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

PRRG2
Western Blot
Unconjugated
Rabbit
Cell Biology
PBS (pH 7.3), 50% glycerol
5639
IgG
Affinity purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human, Mouse, Rat
PRGP2proline-rich Gla (G-carboxglutamic acid) polypeptide 2, proline rich Gla (G-carboxyglutamic acid) 2, Proline-rich gamma-carboxyglutamic acid protein 2, proline-rich Gla (G-carboxyglutamic acid) polypeptide 2, Proline-rich Gla protein 2, TMG2, transmembrane gamma-carboxyglutamic acid protein 2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-179 of human PRRG2 (NP_001303264.1). MSLGPEVFLGPPEAQSFLSSHTRIPRANHWDLELLTPGNLERECLEERCSWEEAREYFEDNTLTERFWESYIYNGKGGRGRVDVASLAVGLTGGILLIVLAGLGAFWYLRWRQHRGQQPCPQEAGLISPLSPLNPLGPPTPLPPPPPPPPGLPTYEQALAASGVHDAPPPPYTSLRRPH
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.