missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRRC2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569520
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.
PRRC2B (Proline Rich Coiled-Coil 2B) is a Protein Coding gene. An important paralog of this gene isPRRC2C.
Specifications
| PRRC2B | |
| Polyclonal | |
| Unconjugated | |
| PRRC2B | |
| 5830434P21Rik; AI173903; BAT2L; Bat2l1; D430039P21; HLA-B associated transcript 2-like 1; HLA-B-associated transcript 2-like 1; Kiaa0515; LQFBS-1; mKIAA0515; proline rich coiled-coil 2B; proline-rich coiled-coil 2B; proline-rich coiled-coil protein 2B; protein BAT2-like 1; protein PRRC2B; PRRC2B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84726 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5JSZ5 | |
| PRRC2B | |
| synthetic peptide directed towards the N terminal region of human PRRC2B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction