missing translation for 'onlineSavingsMsg'
Learn More

PRR7 Antibody (TRAP3/10), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18215961 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215961 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18215961 Supplier Novus Biologicals Supplier No. NBP130424JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRR7 Monoclonal specifically detects PRR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR7
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone TRAP3/10
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias MGC10772, proline rich 7 (synaptic), proline-rich protein 7, Synaptic proline-rich membrane protein
Gene Symbols PRR7
Host Species Mouse
Immunogen Recombinant C-terminal half of the intracellular domain of human PRR7/TRAP3 (amino acids 126-253)
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 80758
Test Specificity The mouse monoclonal antibody TRAP3/10 recognizes an epitope located in the C-terminal part of the intracellular domain of PRR7/TRAP3 (amino acids 126-253 of human PRR7 TRAP3), a 28 kDa proline-rich membrane protein presumably associated with NMDA receptor complex.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.