missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRR5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19968
This item is not returnable.
View return policy
Description
PRR5 Polyclonal specifically detects PRR5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PRR5 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
| FLJ20185, FLJ20185k, proline rich 5 (renal), protein observed with Rictor-1 | |
| Rabbit | |
| 31 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P85299 | |
| PRR5 | |
| Recombinant protein encompassing a sequence within the center region of human Protor 1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 55615 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction