missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRR35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564407
This item is not returnable.
View return policy
Description
Immunogen sequence: EHVGEDLTRA LGDYARVEQR LGQLGPAGGL APRPLREQLG KIRLELLTIH QALEQAVRPP DAPLDLSVKR A Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
Specifications
| PRR35 | |
| Polyclonal | |
| Unconjugated | |
| PRR35 | |
| C16orf11; LA16c-366D1.2; proline rich 35; proline-rich protein 35; PRR35; Uncharacterized protein RJD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 146325 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P0CG20 | |
| PRR35 | |
| Recombinant protein corresponding to Human PRR35. Recombinant protein control fragment (Product #RP-103692). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction