missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRR15L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546599
This item is not returnable.
View return policy
Description
Peptide sequence: RLEKIVDKST KGKHVKVSNS GRFKEKKKVR ATLAENPNLF DDHEEGRSSK Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
| PRR15L | |
| Polyclonal | |
| Unconjugated | |
| PRR15L | |
| ATAD4; ATPase family, AAA domain containing 4; proline rich 15 like; proline rich 15-like; proline-rich protein 15-like protein; Protein ATAD4; PRR15L; testicular tissue protein Li 157 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79170 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BU68 | |
| PRR15L | |
| Synthetic peptide directed towards the C-terminal of human PRR15L (aa 54-103). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction