missing translation for 'onlineSavingsMsg'
Learn More

PRR12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£178.00 - £238.00

Specifications

Antigen PRR12
Dilution Immunohistochemistry-Paraffin 1:200-1:1000
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18680179
View Documents
Novus Biologicals
NBP3-06355-100ul
100 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18670939
View Documents
Novus Biologicals
NBP3-06355-50ul
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

PRR12 Polyclonal antibody specifically detects PRR12 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

PRR12
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
KIAA1205, proline rich 12, proline-rich protein 12
Produced in rabbits immunized with E. coli-derived Human PRR12 fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunohistochemistry-Paraffin 1:200-1:1000
Polyclonal
Purified
RUO
PBS, pH7.0
57479
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.