missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRR11 Antibody (OTI2A10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73677APC
This item is not returnable.
View return policy
Description
PRR11 Monoclonal antibody specifically detects PRR11 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| PRR11 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 55771 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2A10 | |
| Western Blot, Immunohistochemistry | |
| FLJ11029, proline rich 11, proline-rich protein 11, transcription repressor of MHCII | |
| Human recombinant protein fragment corresponding to amino acids 125-360 of human PRR11 (NP_060774) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction