missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PrPSc Monoclonal specifically detects PrPSc in Human, Hamster samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | Prion protein |
| Applications | ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 3F4 |
| Conjugate | FITC |
| Dilution | ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | PBS |
| Gene Alias | CD230, CJD, fatal familial insomnia), GSS, prion protein, prion protein (p27-30), prion protein PrP, prion-related protein, PRIPMGC26679 |
| Gene Symbols | PRNP |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?