missing translation for 'onlineSavingsMsg'
Learn More

PrPSc Antibody (3F4), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18371921 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371921 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371921 Supplier Novus Biologicals Supplier No. NBP281078AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PrPSc Monoclonal specifically detects PrPSc in Human, Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prion protein
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3F4
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD230, CJD, fatal familial insomnia), GSS, prion protein, prion protein (p27-30), prion protein PrP, prion-related protein, PRIPMGC26679
Gene Symbols PRNP
Host Species Mouse
Immunogen Hamster 263K PrPs.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5621
Target Species Human, Hamster
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.