missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PRPSAP2 Monoclonal antibody specifically detects PRPSAP2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | PRPSAP2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI1E3 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | MGC117304, MGC126719, MGC126721, PAP4141 kDa phosphoribosypyrophosphate synthetase-associated protein, phosphoribosyl pyrophosphate synthase-associated protein 2, phosphoribosyl pyrophosphate synthetase-associated protein 2, PRPP synthase-associated protein 2 |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human PRPSAP2 (NP_002758) produced in HEK293T cell. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?