missing translation for 'onlineSavingsMsg'
Learn More

PRPS2 Antibody (4C1), Novus Biologicals™

Product Code. 18326058 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326058 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326058 Supplier Novus Biologicals Supplier No. H00005634M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRPS2 Monoclonal antibody specifically detects PRPS2 in Human samples. It is validated for Western Blot, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPS2
Applications Western Blot, ELISA, KnockDown
Classification Monoclonal
Clone 4C1
Conjugate Unconjugated
Dilution Western Blot 1:500RNAi Validation, ELISA, Knockdown Validated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002756
Gene Alias EC 2.7.6.1, Phosphoribosyl pyrophosphate synthase II, phosphoribosyl pyrophosphate synthetase 2, PPRibP, PPRibP synthetase, PRSII, PRS-II, ribose-phosphate diphosphokinase 2, ribose-phosphate pyrophosphokinase 2
Host Species Mouse
Immunogen PRPS2 (NP_002756, 160 a.a. ~ 269 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AEWKNCIIVSPDAGGAKRVTSIADRLNVEFALIHKERKKANEVDRMVLVGDVKDRVAILVDDMADTCGTICHAADKLLSAGATKVYAILTHGIFSGPAISRINNAAFEAV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5634
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.