missing translation for 'onlineSavingsMsg'
Learn More

PRPS1L1 Antibody [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30763842 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763842 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763842 Supplier Novus Biologicals Supplier No. NBP346878MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRPS1L1 Polyclonal antibody specifically detects PRPS1L1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPS1L1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.6.1, Phosphoribosyl pyrophosphate synthase III, phosphoribosyl pyrophosphate synthetase 1-like 1, phosphoribosylpyrophosphate synthetase subunit III, PRPS1, PRPS1-like 1, PRPS3Phosphoribosyl pyrophosphate synthase 1-like 1, PRPSL, PRS-III, ribose-phosphate diphosphokinase catalytic chain III, ribose-phosphate pyrophosphokinase 3, ribose-phosphate pyrophosphokinase III
Host Species Rabbit
Immunogen phosphoribosyl pyrophosphate synthetase 1-like 1. (Uniprot# P21108)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 221823
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.