missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPS1 Antibody (1E11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42649CL1
This item is not returnable.
View return policy
Description
PRPS1 Monoclonal antibody specifically detects PRPS1 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| PRPS1 | |
| Monoclonal | |
| CoraFluor 1 | |
| CMTX5ARTS, deafness, X-linked 2, perceptive, congenital, DFN2, DFNX1KIAA0967, dJ1070B1.2 (phosphoribosyl pyrophosphate synthetase 1), EC 2.7.6.1, Phosphoribosyl pyrophosphate synthase I, phosphoribosyl pyrophosphate synthetase 1, PPRibP, PRSI, PRS-I, ribose-phosphate diphosphokinase 1, ribose-phosphate pyrophosphokinase 1 | |
| Recombinant human PRPS1 (1-318aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis, Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers | |
| 5631 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 1E11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction