missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRPK Monoclonal Antibody (OTI5G7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526955
This item is not returnable.
View return policy
Description
PRPK Monoclonal Antibody for Western Blot, IHC (P)
Component of the EKC/KEOPS complex that is required for the formation of a threonylcarbamoyl group on adenosine at position 37 (t(6)A37) in tRNAs that read codons beginning with adenine (PubMed:22912744, PubMed:27903914). The complex is probably involved in the transfer of the threonylcarbamoyl moiety of threonylcarbamoyl-AMP (TC-AMP) to the N6 group of A37 (PubMed:22912744, PubMed:27903914). TP53RK has ATPase activity in the context of the EKC/KEOPS complex and likely plays a supporting role to the catalytic subunit OSGEP. Atypical protein kinase that phosphorylates 'Ser-15' of p53/TP53 protein and may therefore participate in its activation (PubMed:11546806). [UniProt].
Specifications
| PRPK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96S44 | |
| Tp53rk | |
| Full length human recombinant protein of TP53RK produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5G7 | |
| Unconjugated | |
| Tp53rk | |
| 2810408M09Rik; 4933401B08Rik; 5630401H01Rik; Atypical serine/threonine protein kinase TP53RK; BUD32; C20orf64; dJ101A2; EKC/KEOPS complex subunit TP53RK; mNori-2p; Nori-2; Nori-2p; p53-related protein kinase; PRPK; TP53 regulating kinase; TP53-regulating kinase; TP53RK; transformation related protein 53 regulating kinase; Trp53 regulating kinase; Trp53rk; Trp53tk | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 112858 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction