missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF8 Rabbit anti-Human, Mouse, Rat, Clone: 4I1Q8, Novus Biologicals™
Rabbit Monoclonal Antibody
£173.00 - £405.00
Specifications
| Antigen | PRPF8 |
|---|---|
| Clone | 4I1Q8 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18316845
|
Novus Biologicals
NBP3-16440-20UL |
20 μg |
£173.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18386786
|
Novus Biologicals
NBP3-16440-100UL |
100 μg |
£405.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRPF8 Monoclonal antibody specifically detects PRPF8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| PRPF8 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| apoptosis-regulated protein 1, apoptosis-regulated protein 2, p220, precursor mRNA processing protein, pre-mRNA-processing-splicing factor 8, Prp8, PRP8 homolog, PRP8 pre-mRNA processing factor 8 homolog (S. cerevisiae), PRP8 pre-mRNA processing factor 8 homolog (yeast), PRP8,220 kDa U5 snRNP-specific protein, PRPC8HPRP8, RP13, Splicing factor Prp8, U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 2200-2335 of human PRPF8 (Q6P2Q9). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 4I1Q8 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Vision | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 10594 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title