missing translation for 'onlineSavingsMsg'
Learn More

PRPF8 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18406425 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406425 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406425 Supplier Novus Biologicals Supplier No. NBP222274DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRPF8 Polyclonal antibody specifically detects PRPF8 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF8
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias apoptosis-regulated protein 1, apoptosis-regulated protein 2, p220, precursor mRNA processing protein, pre-mRNA-processing-splicing factor 8, Prp8, PRP8 homolog, PRP8 pre-mRNA processing factor 8 homolog (S. cerevisiae), PRP8 pre-mRNA processing factor 8 homolog (yeast), PRP8,220 kDa U5 snRNP-specific protein, PRPC8HPRP8, RP13, Splicing factor Prp8, U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 2285 and 2335 of human Pre-mRNA-Processing Factor 8 using the numbering given in entry NP_006436.3 (GeneID 10594).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 10594
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.