missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRPF4B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110223
This item is not returnable.
View return policy
Description
Immunogen sequence: QKYKYLAEDS NMSVPSEPSS PQSSTRTRSP SPDDILERVA ADVKEYEREN VDTFEASVKA KHNLMTVEQN NGSSQKKLLA PDMFTESDDM FAAYFDSARL RAAGIGKDFK ENPNLRDNWT DAEGYYRVNI GEVLDKRYNV YGYTGQGVFS NVVRARDNAR ANQEVAVKII RNNELMQKTG LKELEFLKKL NDADPDDKFH CLRLFRHFYH K.
Pre-mRNA splicing occurs in two sequential transesterification steps, and the protein encoded by this gene is thought to be involved in pre-mRNA splicing and in signal transduction. This protein belongs to a kinase family that includes serine/arginine-rich protein-specific kinases and cyclin-dependent kinases. This protein is regarded as a CDK-like kinase with homology to mitogen-activated protein kinases.
Specifications
| PRPF4B | |
| Polyclonal | |
| Unconjugated | |
| PRPF4B | |
| 2610037H07; cbp143; dJ1013A10.1; dJ1013A10.1 (PRP4 protein kinase homolog); KIAA0536; PR4H; pre-mRNA processing factor 4B; pre-mRNA protein kinase; protein-serine/threonine kinase; Prp4; PRP4 kinase; PRP4 pre-mRNA processing factor 4 homolog B; PRP4 pre-mRNA processing factor 4 homolog B (yeast); PRP4 pre-mRNA-processing factor 4 homolog; PRP4H; PRP4K; Prp4m; Prpf4b; Prpk; RP5-1013A10.2; Serine/threonine-protein kinase PRP4 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19134, 291078, 8899 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.24 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q13523, Q5RKH1, Q61136 | |
| PRPF4B | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction