missing translation for 'onlineSavingsMsg'
Learn More

PRPF4B Antibody (3E10), Novus Biologicals™

Product Code. 18356328 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356328 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356328 Supplier Novus Biologicals Supplier No. H00008899M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRPF4B Monoclonal antibody specifically detects PRPF4B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF4B
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 3E10
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003904
Gene Alias dJ1013A10.1, dJ1013A10.1 (PRP4 protein kinase homolog), KIAA0536EC 2.7.11.1, PR4H, protein-serine/threonine kinase, PRP4 kinase, PRP4 pre-mRNA processing factor 4 homolog B (yeast), PRP4 pre-mRNA-processing factor 4 homolog, PRP4H, PRP4K, PRP4Prp4, serine/threonine-protein kinase PRP4 homolog
Host Species Mouse
Immunogen PRPF4B (NP_003904.2, 898 a.a. ~ 1005 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LKLAMDLKGKMPNKMIRKGVFKDQHFDQNLNFMYIEVDKVTEREKVTVMSTINPTKDLLADLIGCQRLPEDQRKKVHQLKDLLDQILMLDPAKRISINQALQHAFIQE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 8899
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.