missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRPF4 Recombinant Rabbit Monoclonal Antibody (24GB980)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | PRPF4 |
|---|---|
| Clone | 24GB980 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276432
![]() |
Invitrogen™
MA557185 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274965
|
Invitrogen™
MA557186 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRPF4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF4 is 1 of several proteins that associate with U4 and U6 snRNPs.Specifications
| PRPF4 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| PRPF4 | |
| 1600015H11Rik; AI874830; AW047464; bN189G18.1; HPRP4; HPRP4P; hypothetical protein LOC508735; mg:ab03a02; pre-mRNA processing factor 4; PRP4; PRP4 homolog; PRP4 pre-mRNA processing factor 4 homolog; PRP4 pre-mRNA processing factor 4 homolog (yeast); PRP4/STK/WD splicing factor; Prp4p; PRPF4; RP70; SNRNP60; U4/U6 small nuclear ribonucleoprotein Prp4; U4/U6 snRNP 60 kDa protein; WD splicing factor Prp4; zgc:65943 | |
| PRPF4 | |
| Primary | |
| Affinity chromatography |
| 24GB980 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43172, Q9DAW6 | |
| 298095, 70052, 9128 | |
| A synthesized peptide derived from human PRPF4 (aa 480-522). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title