missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRPF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598090
This item is not returnable.
View return policy
Description
PRPF4 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF4 is 1 of several proteins that associate with U4 and U6 snRNPs.
Specifications
| PRPF4 | |
| Polyclonal | |
| Unconjugated | |
| PRPF4 | |
| 1600015H11Rik; AI874830; AW047464; bN189G18.1; HPRP4; HPRP4P; hypothetical protein LOC508735; mg:ab03a02; pre-mRNA processing factor 4; PRP4; PRP4 homolog; PRP4 pre-mRNA processing factor 4 homolog; PRP4 pre-mRNA processing factor 4 homolog (yeast); PRP4/STK/WD splicing factor; Prp4p; PRPF4; RP70; SNRNP60; U4/U6 small nuclear ribonucleoprotein Prp4; U4/U6 snRNP 60 kDa protein; WD splicing factor Prp4; zgc:65943 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9128 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43172 | |
| PRPF4 | |
| Recombinant Human U4/U6 small nuclear ribonucleoprotein Prp4 protein (1-260AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction