Learn More
PRPF38B Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23901-1-AP-20UL
Description
The PRPF38B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PRPF38B. This antibody recognizes human, mouse antigen. The PRPF38B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PRPF38B | |
| 200 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| Pre mRNA splicing factor 38B, PRPF38B, RP11 293A10.1, Sarcoma antigen NY SAR 27 | |
| 20 μL | |
| 55119 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |