missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PrPC Rabbit anti-Human, Mouse, Rat, Clone: 3D0J9, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16708-100UL
This item is not returnable.
View return policy
Description
PrPC Monoclonal antibody specifically detects PrPC in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PrPC | |
| Monoclonal | |
| Unconjugated | |
| CD230, CJD, fatal familial insomnia), GSS, prion protein, prion protein (p27-30), prion protein PrP, prion-related protein, PRIPMGC26679 | |
| A synthetic peptide corresponding to a sequence within amino acids 154-253 of human PrPC (P04156). | |
| 100 μg | |
| Neuroscience, Stem Cell Markers | |
| 5621 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 3D0J9 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction