missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRP8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16589331
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16589331 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16589331 Supplier Invitrogen™ Supplier No. PA596497

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MAGVFPYRGP GNPVPGPLAP LPDYMSEEKL QEKARKWQQL QAKRYAEKRK FGFVDAQKED MPPEHVRKII RDHGDMTNRK FRHDKRVYLG ALKYMPHAVL; Positive Samples: HeLa, Jurkat; Cellular Location: Nucleus speckle.

Pre-mRNA splicing occurs in 2 sequential transesterification steps. The protein encoded by this gene is a component of both U2- and U12-dependent spliceosomes, and found to be essential for the catalytic step II in pre-mRNA splicing process. It contains several WD repeats, which function in protein-protein interactions. This protein has a sequence similarity to yeast Prp8 protein. This gene is a candidate gene for autosomal dominant retinitis pigmentosa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRP8
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Prpf8
Gene Accession No. Q6P2Q9, Q99PV0
Gene Alias 220 kDa U5 snRNP-specific protein; apoptosis-regulated protein 1; apoptosis-regulated protein 2; AU019467; D11Bwg0410e; DBF3/PRP8; H PRPC8; HPRP8; id:ibd1257; ik:tdsubc_2a9; im:7141966; p220; precursor mRNA processing protein; pre-mRNA processing factor 8; pre-mRNA-processing-splicing factor 8; Prp8; PRP8 homolog; PRP8 pre-mRNA processing factor 8 homolog; PRPC8; prpf8; RP13; Sfprp8l; SNRNP220; spl; Splicing factor Prp8; tdsubc_2a9; U5 snRNP-specific protein; U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p; wu:fb37c02; wu:fb73e06; xx:tdsubc_2a9; zgc:56504
Gene Symbols Prpf8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PRPF8 (NP_0064363).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10594, 192159, 287530
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.