missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRP19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529254
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, Molt-4. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (83%), Pig (100%), Chicken (97%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PSO4 is the human homolog of yeast Pso4, a gene essential for cell survival and DNA repair.
Specifications
| PRP19 | |
| Polyclonal | |
| Unconjugated | |
| Prpf19 | |
| AA617263; AL024362; D19Wsu55e; hPSO4; Neuronal differentiation-related gene protein; NMP200; Nuclear matrix protein 200; nuclear matrix protein NMP200 related to splicing factor PRP19; nuclear matrix protein SNEV; pre-mRNA processing factor 19; pre-mRNA-processing factor 19; PRP19; prp19 beta; PRP19/PSO4 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog (S. cerevisiae); PRPF19; PSO4; psoralen 4; RING-type E3 ubiquitin transferase PRP19; Senescence evasion factor; SNEV; UBOX4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27339, 28000 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.29 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q99KP6, Q9UMS4 | |
| Prpf19 | |
| Recombinant protein encompassing a sequence within the center region of human PRP19. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction