missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ PRP™-X300 Ion Exclusion HPLC Columns
Description
A fast, versatile HPLC column packing material, Hamilton PRP-X300 columns rapidly and easily separate closely related alcohols and organic acids without column heating. The sulfonated poly(styrene-divinylbenzene) support separates samples by a mixed mode mechanism.
Three Separation Modes
- Hydrogen Bonding, Reversed Phase, and Ion Exclusion
- A wide range of samples can be analyzed with PRP-X300 columns because the selectivity of the column can be altered by changing the pH of the buffer or adding an organic modifier (Methanol, Acetonitrile) to the buffer. The supports stability to organic solvents makes it possible to analyze samples that have been too highly retained on conventional ion exclusion supports. The use of isocratic conditions allows samples to be analyzed without column reequilibration.
- The excellent selectivity of the PRP-X300 columns ensures most separations are complete within five minutes
- Most samples require only minimal preparation before injection
- One particle size: 7μm
- Three column internal diameters: 1.0 to 4.6mm
- Two column materials: 316 stainless steel and PEEK
- Analytical and semiprep/preparative guard columns
Specifications
Specifications
| Column Format | Analytical Column |
| Diameter (Metric) | 4.1 mm |
| Length (Metric) | 250 mm |
| Max. Pressure | 5000 psi (345 bar) |
| Mobile Phase Compatibility | pH 1 to 13, 0 to 100% aqueous, organic modifier |
| Particle Size | 7 μm |
| pH | 1 to 13 |
| Pore Size | 100 Å |
| Temperature | 5°C to 60°C |
| USP Type | L17 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?