missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PrP Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518539
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
Human prion protein (PrP), also known as PRNP, is a ubiquitously expressed GPI-anchored cell surface glycoprotein associating with lipid raft components and functioning as a signaling molecule. PrP plays a role in apoptosis in a cell context-dependent manner, is involved in proliferation of epithelial cells and in distribution of junction-associated proteins in human enterocytes. Conversion of this normal cellular prion protein (PrPc) into an abnormal conformer (PrPSc) is the crucial step associated with triggering the pathogenesis of the prion neurodegenerative disorders, such as the Creutzfeld-Jakob disease (CJD). Whereas PrPc is rich in alpha-helices, the PrPSc form has higher content of beta-sheets and is resistant to proteinase K.
Specifications
| PrP | |
| Polyclonal | |
| Unconjugated | |
| PRNP | |
| AA960666; AI325101; Alternative prion protein; alternative prion protein; major prion protein; AltPrP; ASCR; BSE; CD230; CD230 antigen; CJD; GSS; KURU; major prion protein; major prion protein; alternative prion protein; major scrapie-associated fibril protein 1; MGC26679; p27-30; PK resistant core; prion protein; prion protein (p27-30) (Creutzfeldt-Jakob disease, Gerstmann-Strausler-Scheinker syndrome, fatal familial insomnia); prion protein precursor; prion protein precursor PrP; prion protein PrP; prion protein variant a; prion protein variant b; prion protein, PrP; prion protein, structural; prion-related protein; PRIP; Prn; Prn-i; Prnp; Prn-p; PrP; PrP>C>; PrP27-30; PrP33-35C; PrPc; PrPSc; Sinc; SIP; TSE | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 5621 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| F7VJQ1, P04156 | |
| PRNP | |
| Synthetic peptide sequence (SDYEDRYYREN) corresponding to the internal amino acids of PRNP (aa 143-153). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction