Learn More
Hamilton™ PRP-1 Reversed-Phase HPLC Column, 100 x 250 mm, 12-20 μm, Repack
PRP-1 columns have inherent reversed-phase separation qualities that eliminate the requirement for a stationary phase coating. Hydrolysis of the stationary phase coating is often the limiting factor in column lifetime, so the lack of coating on the PRP-1 resin extends the longevity of the column. PRP-1 also have exceptional sample recovery (≥ 95%).
Brand: Hamilton™ 79800
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Specification Sheet
Separation Mode | Reversed-Phase |
---|---|
Stationary Phase | PRP-1 |
Format | Column Repack |
Hardware Inner Diameter | 100 mm 101.6 mm |
Hardware Length | 250 mm |
Particle Size | 12-20 μm |
Packing Material Type | PSDVB |
Pore Size | 100 Å |
Hardware Material | Stainless Steel |
Max Pressure | 2,000 psi |
Mobile Phase Limits | pH 1-13. 0-100% aqueous, organic modifier |
Buffer Strength | 0.0-0.5N |
Temp Limits | 5-85°C |
Restoration | Run a gradient of 100% water to 100% acetonitrile. Repeat three times |
Compound Classification | Organics |
UOM | 1 EACH |
Specifications
12 to 20 μm | |
PSDVB | |
Hardware material- Stainless Steel | |
PRP-1 Reversed-Phase HPLC Column |