missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PRP-1 5 μm 100 Å 10.0 x 100 mm

Product Code. 11588124
Change view
Click to view available options
Length (Metric):
100 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11588124 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11588124 Supplier Hamilton™ Supplier No. 79355
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PRP-1 columns have inherent reversed-phase separation qualities that eliminate the requirement for a stationary phase coating. Hydrolysis of the stationary phase coating is often the limiting factor in column lifetime, so the lack of coating on the PRP-1 resin extends the longevity of the column. PRP-1 also have exceptional sample recovery (≥ 95%).

Specification Sheet

  • Separation Mode – Reversed-Phase
  • Stationary Phase – PRP-1
  • Format – HPLC Column
  • Hardware Inner Diameter – 10 mm
  • Hardware Length – 100 mm
  • Particle Size – 5 μm
  • USP Classification – USP L21
  • Packing Material Type – PSDVB
  • Pore Size – 100 Å
  • Hardware Material – Stainless Steel
  • Max Pressure – 5,000 psi
  • Mobile Phase Limits – pH 1-13. 0-100% aqueous, organic modifier
  • Buffer Strength – 0.0-0.5N
  • Temp Limits – 5-85°C
  • Restoration – Run a gradient of 100% water to 100% acetonitrile. Repeat three times
  • Compound Classification – Organics
  • UOM – 1 EACH

Specifications

Column Format HPLC Column
Diameter (Metric) 10 mm
Length (Metric) 100 mm
Max. Pressure 5,000 psi
Particle Size 5 μm
pH pH 1-13. 0-100% aqueous, organic modifier
Pore Size 100 Å
Temperature 5° to 85°C
USP Type USP L21
Packing Material PSDVB
Phase Reversed Phase
Material Stainless Steel
Product Type PRP-1 Columns
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.