missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ PRP-1 5 μm 100 Å 1.0 x 150 mm
Click to view available options
Length (Metric):
150 mm
Unit Size:
Each
Description
Specification Sheet
- Separation Mode – Reversed-Phase
- Stationary Phase – PRP-1
- Format – HPLC Column
- Hardware Inner Diameter – 1.0 mm
- Hardware Length – 150 mm
- Particle Size – 5 μm
- USP Classification – USP L21
- Packing Material Type – PSDVB
- Pore Size – 100 Å
- Hardware Material – Stainless Steel
- Max Pressure – 5,000 psi
- Mobile Phase Limits – pH 1-13. 0-100% aqueous, organic modifier
- Buffer Strength – 0.0-0.5N
- Temp Limits – 25-90°C
- Restoration – Run a gradient of 100% water to 100% acetonitrile. Repeat three times
- Compound Classification – Organics
- UOM – 1 EACH
Specifications
Specifications
| Column Format | HPLC Column |
| Diameter (Metric) | 1.0 mm |
| Length (Metric) | 150 mm |
| Max. Pressure | 5,000 psi |
| Particle Size | 5 μm |
| pH | pH 1-13. 0-100% aqueous, organic modifier |
| Pore Size | 100 Å |
| Temperature | 25°C to 90°C |
| USP Type | USP L21 |
| Packing Material | PSDVB |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction